Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentanone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018514
Click to view available options
Quantity:
5 mL
50 mL

Cyclopentanone is one of the industrially important compounds employed in the synthesis of fungicides, pharmaceuticals, rubber chemicals and perfume products.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 120-92-3
Density 0.951 g/mL (at 25°C)
Flash Point 30°C - Closed Cup
Linear Formula C5H8(=O)
MDL Number MFCD00001409
Melting Point -51°C
Boiling Point 130°C to 131°C
Type Analytical Standard
Quantity 50 mL
Formula Weight 84.12
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.