Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentene, 98+%

Catalog No. p-7051534
Click to view available options
Quantity:
25 mL
100 mL

CAS: 142-29-0 | C5H8 | 68.119 g/mol

Cyclopentene is used in organic synthesis for cross-linking resin. It is used as an intermediate in Industries such as Agrochemical, Dyestuff, Pharmaceutical, syntheses material, Chemical. It is used as a monomer for synthesis of plastics and rubber. It is also used for the synthesis of various chemicals, such as 1,1,2-Trimethylcyclohexane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135°C
Color Colorless
Density 0.773 g/mL
Boiling Point 44°C to 45°C
Flash Point -29°C (-20°F)
Refractive Index 1.4225
Quantity 25 mL
UN Number UN2246
Beilstein 635707
Sensitivity Air Sensitive
Solubility Information It is soluble in most organic solvents like ethanol and ether. It is immiscible with water.
Formula Weight 68.12
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Cyclopentene
Show More Show Less
Hazard Category H225-H302+H312-H304-H315-H319
Hazard Statement GHS H Statement
H225-H304-H302-H312-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Harmful in contact with skin.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P312-P330-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOPENTENE
EINECSNumber 205-532-9
RTECSNumber GY5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.