Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentenylferrocene 96.0+%, TCI America™
SDP

Catalog No. F031325G
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 12260-67-2
Molecular Formula C15H16Fe
MDL Number MFCD00191436
Synonym Cyclopentenylbis(cyclopentadienyl) Iron, Ferrocenecyclopenten-1-yl

Specifications

Melting Point 66°C
Quantity 25 g
Formula Weight 252.14
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentenylferrocene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.