Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentylacetic acid, 97%

Catalog No. p-97433
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

Catalog No. AC154870100

Supplier: Thermo Scientific Chemicals 154870100

Only null left
Add to Cart

CAS: 1123-00-8 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-00-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001387
InChI Key YVHAIVPPUIZFBA-UHFFFAOYSA-N
Synonym cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
PubChem CID 71606
IUPAC Name 2-cyclopentylacetic acid
SMILES OC(=O)CC1CCCC1

Specifications

Color Colorless
Density 1.0200g/mL
Boiling Point 133.0°C to 134.0°C (23.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Linear Formula C5H9CH2CO2H
Quantity 10 g
Beilstein 09, 10
Specific Gravity 1.02
Formula Weight 128.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclopentylacetic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-368-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.