Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclophilin 40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119062
This item is not returnable.
View return policy
Description
Cyclophilin 40 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
Specifications
Cyclophilin 40 | |
Polyclonal | |
Unconjugated | |
PPID | |
40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
361967, 5481, 67738 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q08752, Q6DGG0, Q9CR16 | |
PPID | |
Recombinant protein Cyclophilin 40. The antigen corresponds to amino acid range 7-191 of the target protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction