Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclophilin 40 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5122054

 View more versions of this product

Catalog No. PIPA5122054


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Cyclophilin 40 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclophilin 40
Polyclonal
Unconjugated
PPID
40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147
Rabbit
Antigen affinity chromatography, Protein A
RUO
361967, 5481, 67738
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q08752, Q6DGG0, Q9CR16
PPID
Recombinant protein Cyclophilin 40. The antigen corresponds to amino acid range 7-191 of the target protein.
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.