Learn More
Invitrogen™ Cyclophilin 40 Polyclonal Antibody

Description
PA3-023 detects cyclophilin 40 (CyP-40) from human, mouse and rat tissues and cells. PA3-023 has been successfully used in Western blot and gel shift procedures. By Western blot, this antibody detects a 40 kDa protein representing CyP-40 from rat brain extract. The PA3-023 immunogen is recombinant human CyP-40 expressed in E. coli.
Specifications
Specifications
| Antigen | Cyclophilin 40 |
| Applications | Gel Shift, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum, PBS with 0.05% sodium azide |
| Gene | PPID |
| Gene Accession No. | Q08752, Q6DGG0, Q9CR16 |
| Gene Alias | 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147 |
| Gene Symbols | PPID |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.