Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclophilin 40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527988 Supplier Invitrogen™ Supplier No. PA527988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (94%), Pig (91%), Rhesus Monkey (97%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin 40
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PPID
Gene Accession No. Q08752, Q9CR16
Gene Alias 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147
Gene Symbols PPID
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cyclophilin 40. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100752921, 5481, 67738
Target Species Hamster, Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.