Learn More
Invitrogen™ Cyclophilin 40 Recombinant Rabbit Monoclonal Antibody (24GB1560)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA557559

Description
Cyclophilin 40 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
Specifications
Cyclophilin 40 | |
Recombinant Monoclonal | |
Unconjugated | |
PPID | |
40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; 4930564J03Rik; cyclophilin 40; cyclophilin D; cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP40; CYP-40; CYPD; CYP-S1; cytoplasmic cyclophilin D; EC 5.2.1.8; I79_011990; MGC33096; peptidyl-prolyl cis-trans isomerase D; peptidylprolyl isomerase D; peptidylprolyl isomerase D (cyclophilin D); PPIase; PPIase D; PPID; Ppidl; Ppif; rotamase; Rotamase D; S-cyclophilin; SCYLP; testicular tissue protein Li 147 | |
Rabbit | |
Affinity chromatography | |
RUO | |
361967, 5481, 67738 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
24GB1560 | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q08752, Q6DGG0, Q9CR16 | |
PPID | |
A synthesized peptide derived from human Cyclophilin 40 (aa 300-370). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.