Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin A Antibody (001), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290520R
Description
Cyclophilin A Monoclonal specifically detects Cyclophilin A in Mouse samples. It is validated for ELISA.Specifications
Cyclophilin A | |
Monoclonal | |
DyLight 550 | |
Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cyclophilin A (NP_032933.1; Met 1-Leu 164) . | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Stem Cell Markers | |
5478 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction