Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophilin A/PPIA Rat anti-Human, Clone: 8H7, MilliporeSigma™

Catalog No. MABE1924100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1924100 100 μg
MABE192425 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE1924100 Supplier MilliporeSigma Supplier No. MABE1924100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cyclophilin A/PPIA Monoclonal antibody specifically detects Cyclophilin A/PPIA in Human samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assay, Neutralizing, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin A/PPIA
Applications ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Neutralization, Western Blot
Classification Monoclonal
Clone 8H7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in PBS without preservatives.
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Cyclophilin A.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.