Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin B Antibody [DyLight 680], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF5410Y
Description
Cyclophilin B Polyclonal antibody specifically detects Cyclophilin B in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-OutSpecifications
Cyclophilin B | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Gene Knock-Out | |
DyLight 680 | |
Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP | |
E. coli-derived recombinant human Cyclophilin B, Asp34-Glu216, Accession # P23284 | |
0.1 mL | |
Loading Controls | |
5479 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction