Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclophilin B Monoclonal Antibody (CL3901)
GREENER_CHOICE

Catalog No. PIMA531392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531392 Supplier Invitrogen™ Supplier No. MA531392
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: TKTDSRDKPL KDVIIADCGK IEVEKPFAIA KE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 97%.

Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins which bind FK506 are termed FK506 Binding Proteins (FKBP's) and those which bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation. Thus, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL3901
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIB
Gene Accession No. P23284, P24368, P24369
Gene Alias AA408962; AA553318; AI844835; Cphn2; Cphn-2; cyclophilin B; cyclophilin-like protein; Cyp B; CyP-20b; CYPB; CYP-S1; EC 5.2.1.8; epididymis secretory protein Li 39; HEL-S-39; OI9; Peptidyl-prolyl cis-trans isomerase B; peptidylprolyl isomerase B; peptidylprolyl isomerase B (cyclophilin B); PPIase; PPIase B; Ppib; Rotamase; Rotamase B; S-cyclophilin; S-cyclophylin; SCYLP
Gene Symbols PPIB
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human PPIB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19035, 5479, 64367
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.