Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin C Antibody (3C6), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197946AF647
Description
Cyclophilin C Monoclonal specifically detects Cyclophilin C in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Cyclophilin C | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPIC | |
| Recombinant human PPIC (39-197aa) protein purified from E. coli | |
| 100 μL | |
| Primary | |
| Anti-human PPIC mAb, clone 3C6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPIC protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3C6 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| Cyclophilin C, CYPCMGC3673, EC 5.2.1.8, parvulin, peptidyl-prolyl cis-trans isomerase C, peptidylprolyl isomerase C (cyclophilin C), PPIase C, Rotamase C | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 5480 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction