Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin C Antibody (3C6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197946PE
Description
Cyclophilin C Monoclonal specifically detects Cyclophilin C in Human samples. It is validated for Flow Cytometry.Specifications
Cyclophilin C | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PPIC | |
Recombinant human PPIC (39-197aa) protein purified from E. coli | |
100 μL | |
Primary | |
Anti-human PPIC mAb, clone 3C6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPIC protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
3C6 | |
Flow Cytometry | |
Cyclophilin C, CYPCMGC3673, EC 5.2.1.8, parvulin, peptidyl-prolyl cis-trans isomerase C, peptidylprolyl isomerase C (cyclophilin C), PPIase C, Rotamase C | |
Mouse | |
Protein G purified | |
RUO | |
5480 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction