Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cyclophilin C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558679
Description
Immunogen sequence: VFGKVIDGMT VVHSIELQAT DGHDRPLTNC SIINSGKIDV KTPFVVEIAD W Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.
Specifications
Cyclophilin C | |
Polyclonal | |
Unconjugated | |
Ppic | |
Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P45877 | |
Ppic | |
Recombinant protein corresponding to Human Cyclophilin C. Recombinant protein control fragment (Product #RP-96925). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction