Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin C Polyclonal Antibody
Cyclophilin C Polyclonal Antibody
Supplier: Thermo Scientific PA569299
Description
Cyclophilin C Polyclonal Antibody for Western Blot

Specifications
Cyclophilin C | |
Polyclonal | |
Unconjugated | |
Ppic | |
Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19038 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P30412 | |
Ppic | |
synthetic peptide directed towards the middle region of human Cyclophilin C (aa 35-84). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction