Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclophilin C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597261 Supplier Invitrogen™ Supplier No. PA597261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSGAEGFRKR GPSVTAKVFF DVRIGDKDVG RIVIGLFGKV VPKTVENFVA LATGEKGYGY KGSKFHRVIK DFMIQGGDIT TGDGTGGVSI YGETFPDENF KLKHYGIGWV SMANAGPDTN GSQFFITLTK PTWLDGKHVV FGKVIDGMTV VHSIELQATD GHDRPLTNCS IINSGKIDVK TPFVVEIADW; Positive Samples: HeLa, MCF-7, SW480, HepG2; Cellular Location: Cytoplasm.

PPIC is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. Similar to other PPIases, this protein can bind immunosuppressant cyclosporin A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ppic
Gene Accession No. P45877
Gene Alias Cyclophilin C; CyP-20c; CYPC; parvulin; peptidyl-prolyl cis-trans isomerase C; peptidylprolyl isomerase C; peptidylprolyl isomerase C (cyclophilin C); PPIase C; Ppic; Rotamase C
Gene Symbols Ppic
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 23-212 of human PPIC (NP_0009341).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5480
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.