Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin-E Antibody (17E8), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259502APC
Description
Cyclophilin-E Monoclonal antibody specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot, ELISASpecifications
| Cyclophilin-E | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 17E8 | |
| Western Blot, ELISA | |
| Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E | |
| Recombinant human Cyclophilin-E (1-301aa) purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 10450 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction