Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclophilin-E Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299016MFV450
Description
Cyclophilin-E Polyclonal antibody specifically detects Cyclophilin-E in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Cyclophilin-E | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
Cyclophilin E, Cyclophilin-33, CyP-33, CYP33, EC 5.2.1.8, MGC111222, MGC3736, peptidyl-prolyl cis-trans isomerase E, peptidylprolyl isomerase E (cyclophilin E), PPIase E, Rotamase E | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Cyclophilin-E. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
10450 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction