Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclophilin E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564978 Supplier Invitrogen™ Supplier No. PA564978
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGSEPPKAETQ EGEPIAKKAR SNPQVYMDIK IGNKPAGRIQ MLLRSDVVPM TAENFRCLCT HEKGFGFKGS Highest antigen sequence identity to the following orthologs - mouse 96%, rat 94%.

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein contains a highly conserved cyclophilin (CYP) domain as well as an RNA-binding domain. It was shown to possess PPIase and protein folding activities and also exhibit RNA-binding activity. Three alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclophilin E
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIE
Gene Accession No. Q9UNP9
Gene Alias 2010010D16Rik; Ab1-210; Cyclophilin E; cyclophilin-33; CYP33; CYP-33; Peptidyl-prolyl cis-trans isomerase E; peptidylprolyl isomerase E; peptidylprolyl isomerase E (cyclophilin E); PPIase E; PPIE; Rotamase E
Gene Symbols PPIE
Host Species Rabbit
Immunogen Recombinant Human Cyclophilin E. Recombinant protein control fragment (Product #RP-105746).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10450
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.