Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclophosphamide monohydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
6055-19-2
C7H15Cl2N2O2P·H2O
279.09
MFCD00149395
PWOQRKCAHTVFLB-UHFFFAOYSA-N
cyclophosphamide monohydrate,cyclophosphamide hydrate,cytoxan,endoxon,endoxan monohydrate,ciclophosphamide hydrat,cytoxan hydrate,endoxan a,cyclophosphamide hydrated
22420
CHEBI:4026
N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate
C1CNP(=O)(OC1)N(CCCl)CCCl.O
6055-19-2
C7H15Cl2N2O2P·H2O
279.09
MFCD00149395
PWOQRKCAHTVFLB-UHFFFAOYSA-N
cyclophosphamide monohydrate,cyclophosphamide hydrate,cytoxan,endoxon,endoxan monohydrate,ciclophosphamide hydrat,cytoxan hydrate,endoxan a,cyclophosphamide hydrated
22420
CHEBI:4026
N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate
C1CNP(=O)(OC1)N(CCCl)CCCl.O
Packaging Glass bottle
Melting Point 49.0°C to 53.0°C
Quantity 1g
Color White
Flash Point >110°C
Water 6 to 7%
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ELSD) (HPLC)
Merck Index 15, 2743
Solubility Information Solubility in water: 40g/L. Other solubilities: slightly soluble in alcohol,benzene,ethylene,glycol,carbon tetrachloride and dioxane,sparingly soluble in ether and acetone
Formula Weight 279.09
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material Cyclophosphamide monohydrate
Show More Show Less

GHS H Statement
May cause genetic defects.
May cause cancer.
May damage the unborn child.
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Wash face,hands and any exposed skin thoroughly after

GHS Signal Word: Danger