Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cyclophosphamide
GREENER_CHOICESDP

Catalog No. 409150
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-915-0 Supplier Tocris Bioscience™ Supplier No. 4091/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alkylating agent; chemotherapeutic

Nitrogen mustard alkylating agent and prodrug. Phosphoramide mustard (active metabolite) forms DNA cross-links leading to cell death. Inhibits aldehyde dehydrogenase 1 (ALDH1) through its degradation product acrolein. Chemotherapeutic for the treatment of breast cancer; regulates Bax and Bcl-2 expression when administered with etoposide (Cat. No. 1226) in breast cancer cell lines.

Chemical Identifiers

CAS 50-18-0
Molecular Formula C7H15Cl2N2O2P

Specifications

Quantity 50 mg
Formula Weight 261.09
Purity >99%
Molecular Weight (g/mol) 261.083
Chemical Name or Material Cyclophosphamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.