Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanecarbonitrile, 98%

Catalog No. p-7081222
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 5500-21-0 | C4H5N | 67.091 g/mol

Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5500-21-0
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001269
InChI Key AUQDITHEDVOTCU-UHFFFAOYSA-N
Synonym cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
PubChem CID 79637
IUPAC Name cyclopropanecarbonitrile
SMILES C1CC1C#N

Specifications

Density 0.905
Boiling Point 134°C to 135°C
Flash Point 32°C (89°F)
Odor Recognizable
Refractive Index 1.421
Quantity 10 g
UN Number UN3275
Beilstein 635790
Solubility Information Miscible with water.
Formula Weight 67.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarbonitrile
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 226-836-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.