Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropanecarboxaldehyde, 98%

Catalog No. p-2691281
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 1489-69-6 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-69-6
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00012261
InChI Key JMYVMOUINOAAPA-UHFFFAOYSA-N
Synonym cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
PubChem CID 123114
IUPAC Name cyclopropanecarbaldehyde
SMILES C1CC1C=O

Specifications

Density 0.9380g/mL
Boiling Point 98.0°C to 101.0°C
Flash Point 7°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4285 to 1.4305
Linear Formula C3H5CHO
Quantity 1 g
Specific Gravity 0.938
Solubility Information Solubility in water: insoluble.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.