Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropyl(2-thienyl)methanone, 97%, Thermo Scientific™

Catalog No. ac111740050
Click to view available options
:
5g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6193-47-1
CAS Min % 96.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C8H8OS
Refractive Index 1.5835 to 1.5855 (20°C, 589nm)
Quantity 5g
Synonym cyclopropyl 2-thienyl ketone, cyclopropyl thiophen-2-yl methanone, cyclopropyl 2-thienyl methanone, cyclopropyl2-thienylketone, acmc-1b3oh, 2-thienylcyclopropyl ketone, cyclopropyl 2-thienylketone, bidd:gt0674, methanone,cyclopropyl-2-thienyl, pjdfnfstscakpc-uhfffaoysa
InChI Key PJDFNFSTSCAKPC-UHFFFAOYSA-N
SMILES C1CC1C(=O)C2=CC=CS2
IUPAC Name cyclopropyl(thiophen-2-yl)methanone
Molecular Weight (g/mol) 152.211
PubChem CID 80328
Formula Weight 152.22
Percent Purity ≥98.5%
Chemical Name or Material Cyclopropyl 2-thienyl ketone
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.