Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylacetonitrile, 97%, Thermo Scientific™

Catalog No. ac435000050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 6542-60-5
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.12
InChI Key FAUQRRGKJKMEIW-UHFFFAOYSA-N
Synonym cyclopropylacetonitrile, cyclopropaneacetonitrile, 2-cyclopropylethanenitrile, cyclopropyl acetonitrile, cyclopropyl-acetonitrile, acmc-20albr, cyanomethyl cyclopropane, ksc352m6t
PubChem CID 643447
IUPAC Name 2-cyclopropylacetonitrile
SMILES C1CC1CC#N

Specifications

Color Colorless to Yellow
Density 0.8780g/mL
Boiling Point 142°C to 144°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4225 to 1.4245 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.878
Formula Weight 81.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclopropylacetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.