Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cyclopropylacetylene, 97%

Manufacturer:  Alfa Aesar™ H5344006

Quantity 5g

 View more versions of this product

Catalog No. AAH5344006


  Check Availability
Add to cart

Specifications

Description

Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

6746-94-7
66.1
UN3295
Cyclopropylacetylene
C5H6
MFCD02181090
Ethynylcyclopropane

Specifications

−17°C (−1°F)
5g
C5H6
MFCD02181090
Ethynylcyclopropane
97%
51°C
6746-94-7
66.1
UN3295
Cyclopropylacetylene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H318-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit