Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylboronic acid, tech. 85%, Thermo Scientific Chemicals

Catalog Number p-7028843
Click to view available options
Quantity:
1 g
5 g
411235-57-9
C3H7BO2
85.90
MFCD04038750
WLVKDFJTYKELLQ-UHFFFAOYSA-N
cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid
2760897
cyclopropylboronic acid
OB(O)C1CC1
411235-57-9
C3H7BO2
85.90
MFCD04038750
WLVKDFJTYKELLQ-UHFFFAOYSA-N
cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid
2760897
cyclopropylboronic acid
OB(O)C1CC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90°C to 95°C
Quantity 1 g
Formula Weight 85.9
Percent Purity 85%
Grade Technical
Chemical Name or Material Cyclopropylboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit