Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylmagnesium bromide, 0.5M in MeTHF

Catalog No. AAH51158AD
Click to view available options
Quantity:
50 mL
100 mL

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol

Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
Reacts with water.

Notes
Air and Moisture Sensitive, store away from air and water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
SMILES Br[Mg]C1CC1

Specifications

Quantity 50 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
IUPAC Name magnesium;cyclopropane;bromide
Formula Weight 145.28
Concentration or Composition (by Analyte or Components) 0.5M in MeTHF
Chemical Name or Material Cyclopropylmagnesium bromide
Hazard Category H225-H260-H302-H314-H335
Hazard Statement GHS H Statement
H225-H261-H314-H318
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.3; Packaging Group: I
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.