Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™

Catalog No. p-4813442
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 23719-80-4 | C3H5BrMg | 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4, 96-47-9
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
IUPAC Name magnesium;cyclopropane;bromide
SMILES Br[Mg]C1CC1

Specifications

Flash Point −11°C
Packaging AcroSeal™ Glass bottle
Linear Formula C3H5MgBr
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 145.28
Chemical Name or Material Cyclopropylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes serious eye damage.
Causes serious eye irritation.
May cause respiratory irritation.<b
Precautionary Statement GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye prot

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.