Learn More
Cyclopropyltriphenylphosphonium bromide, 98%
Description
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
Specifications
| Melting Point | 183°C to 186°C |
| Quantity | 5 g |
| Beilstein | 6413483 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 383.27 |
| Percent Purity | 98% |
| Chemical Name or Material | Cyclopropyltriphenylphosphonium bromide |
Safety and Handling
| Hazard Category | H312+H332 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c |
| EINECSNumber | 237-970-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.