Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopropylzinc bromide, 0.5M solution in THF, AcroSeal™

Catalog No. ac432820500
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 126403-68-7 | C3H5BrZn | 186.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropylzinc bromide
CAS 126403-68-7, 109-99-9
Color White to Yellow
Density 0.9690g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C3H5BrZn
Quantity 50 mL
MDL Number MFCD08741505
Specific Gravity 0.969
Molecular Weight (g/mol) 186.37
Formula Weight 186.37
Physical Form Powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.