Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclosporin A Antibody (CSZ22), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102569DL594
Description
Cyclosporin A Monoclonal antibody specifically detects Cyclosporin A in Nonspecies specific samples. It is validated for ELISASpecifications
Cyclosporin A | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
CSZ22 | |
ELISA | |
C62H111N11O12;Ciclosporin;Cyclosporin;Cyclosporine;Cyclosporine A | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cyclosporin A conjugated with BSA. | |
0.2 mL | |
Stem Cell Markers | |
Non-species specific | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction