Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclosporin A Antibody (CSZ22), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1102569IR
Description
Cyclosporin A Monoclonal specifically detects Cyclosporin A in Non-species specific samples. It is validated for ELISA.Specifications
Cyclosporin A | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPIA | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Cyclosporin A conjugated with BSA. | |
0.2 mL | |
Cancer, Cell Cycle and Replication, Stem Cell Markers | |
5478 | |
Non-species specific | |
Purified |
ELISA | |
CSZ22 | |
ELISA | |
Cyclophilin A, Cyclosporin A-binding protein, CYPAMGC12404, CYPH, EC 5.2.1.8, MGC117158, MGC23397, peptidyl-prolyl cis-trans isomerase A, peptidylprolyl isomerase A (cyclophilin A), PPIase A, Rotamase A, T cell cyclophilin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This recognises Cyclophilin A in buffer solutions and whole blood extracts with Kd = 10^-8 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction