Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cyclosporin A Monoclonal Antibody (CSZ.22)
GREENER_CHOICE

Catalog No. PIMA191110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191110 Supplier Invitrogen™ Supplier No. MA191110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-91110 detects Cyclosporin A from various samples.

Cyclosporine is a natural cyclic polypeptide immunosuppressant isolated from the fungus Beauveria nivea. The exact mechanism of action of cyclosporine is not known but may involve binding to the cellular protein cytophilin, resulting in inhibition of the enzyme calcineurin. This agent appears to specifically and reversibly inhibit immunocompetent lymphocytes in the G0-or G1-phase of the cell cycle. T-lymphocytes are preferentially inhibited with T-helper cells as the primary target. Cyclosporine also inhibits lymphokine production and release. [National Center for Biotechnology Information. PubChem Compound Database; CID=5284373].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclosporin A
Applications ELISA
Classification Monoclonal
Clone CSZ.22
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C62H111N11O12; Ciclosporin; Cyclophilin A; Cyclosporin; Cyclosporin A-binding protein; Cyclosporine; Cyclosporine A; CYPA; CYPH; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase A; rotamase A; T cell cyclophilin
Host Species Mouse
Immunogen Proprietary immunogen.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.