Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYCS Rabbit anti-Human, Unconjugated, Clone: CYCS/3128R, Abnova™

Catalog No. 89339582 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-582 Supplier Abnova Corporation Supplier No. RAB00603
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CYCS Monoclonal antibody specifically detects CYCS in Dog, Frog, Fruitfly, Horse, Human, Mouse, Pigeon, Rat samples. It is validated for WB-Ti, IHC-P, Flow Cyt.

Specifications

Antigen CYCS
Applications Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone CYCS/3128R
Conjugate Unconjugated
Description cytochrome c, somatic
Gene CYCS
Gene Alias CYC, HCS
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CYCS
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 54205
Target Species Dog, Frog, Fruitfly, Equine, Human, Mouse, Pigeon, Rat
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.