Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva CyDye™ DIFluors: Saturation Dye Labeling Kits
GSA_VA

Catalog No. p-4906596
Click to view available options
Quantity:
100 nM each of Cy3 and Cy5

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

Contain 100nm each Cy3 and Cy5 saturation dyes sufficient for at least 12 labeling reactions, using as little as 5μg protein per reaction. 25-8009-84 contains an additional vial of Cy3 saturation dye (400nm) for labeling up to 500μg of protein in a preparative gel.

For applications where only small amounts of sample are available, for example, in Laser Capture Microdissection.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
For Use With (Application) The Scarce Sample Labeling Kit are designed to be used for applications only small amounts of sample are available, for example, in laser capture microdissection
Quantity 100 nM each of Cy3 and Cy5
Product Type Amersham CyDye DIGE Fluor Labeling Kit
Kit Contents 100nM CyDye DIGE Fluor Cy3 saturation dye for analytical labeling, 100nM CyDye DIGE Fluor Cy5 saturation dye for analytical labeling
Content And Storage -15°C to -30°C
Color Green, Red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.