Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYFIP1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150028B
Description
CYFIP1 Polyclonal antibody specifically detects CYFIP1 in Human samples. It is validated for Western BlotSpecifications
CYFIP1 | |
Polyclonal | |
Western Blot | |
cytoplasmic FMR1 interacting protein 1, cytoplasmic FMR1-interacting protein 1, FLJ45151, KIAA0068cytoplasmic FMRP interacting protein 1, P140SRA-1, selective hybridizing clone, SHYC, Specifically Rac1-associated protein 1, Sra-1, SRA1 | |
The immunogen for this product maps to a region between residue 1203 and 1253 of human Cytoplasmic FMR1 Interacting Protein 1 using the numbering given in entry NP_055423.1 (GeneID 23191). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23191 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction