Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYFIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587780 Supplier Invitrogen™ Supplier No. PA587780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAQVTLEDA LSNVDLLEEL PLPDQQPCIE PPPSSLLYQP NFNTNFEDRN AFVTGIARYI EQATVHSSMN EMLEEGQEYA VMLYTWRSCS RAIPQVKCNE QPNRVEIYEK TVEVLEPEVT KLMNFMYFQR NAIERFCGEV RRLCHAERRK DFVSEAYLIT LGKFINMFAV LDELKNMKCS VKNDHSAYKR AAQFLRKMAD PQSIQESQNL SMFLANHNKI TQSLQQQLEV ISGYEELLAD IVNLCVDYYE NRMYLTPSEK HMLLKVMGFG; Positive Samples: B cells, HepG2, HT-1080, Mouse lung, Mouse brain; Cellular Location: Cell junction, Cell projection, Cytoplasm, lamellipodium, perinuclear region, ruffle, synapse, synaptosome.

Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit is an adapter between EIF4E and FMR1. Promotes the translation repression activity of FMR1 in brain probably by mediating its association with EIF4E and mRNA. Involved in formation of membrane ruffles and lamellipodia protrusions and in axon outgrowth. Binds to F-actin but not to RNA. Regulator of epithelial morphogenesis. May act as an invasion suppressor in cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYFIP1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cyfip1
Gene Accession No. Q7L576, Q7TMB8
Gene Alias CY; CYFIP; cyfip1; cyfip1 protein; cytoplasmic FMR1 interacting protein 1; cytoplasmic FMR1-interacting protein 1; Cytoplasmic FMR1-interacting protein 1 homolog; cytoplasmic FMRP interacting protein 1; E030028J09Rik; fb15a06; FLJ45151; FMR1-interacting protein 1; KIAA0068; l(7)1Rl; l71Rl; l7Rl1; LOW QUALITY PROTEIN: cytoplasmic FMR1-interacting protein 1; mKIAA0068; P140sra1; p140sra-1; pl-1; selective hybridizing clone; Shyc; Specifically Rac1-associated protein 1; Sra1; Sra-1; Unknown (protein for IMAGE:7152255); wu:fb15a06
Gene Symbols Cyfip1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CYFIP1 (NP_0554231).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20430, 23191
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.