Learn More
CYFIP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302996AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit is an adapter between EIF4E and FMR1. Promotes the translation repression activity of FMR1 in brain probably by mediating its association with EIF4E and mRNA (By similarity). Involved in formation of membrane ruffles and lamellipodia protrusions and in axon outgrowth. Binds to F-actin but not to RNA. Regulator of epithelial morphogenesis. May act as an invasion suppressor in cancers.Specifications
CYFIP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7L576 | |
CYFIP1 | |
Between 1203 and 1253 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CYFIP1 | |
P140SRA-1, SHYC, SRA1, cytoplasmic FMR1-interacting protein 1, cytoplasmic FMRP interacting protein 1, selective hybridizing clone, specifically Rac1-associated protein 1, sra-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23191 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.