Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYFIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567174 Supplier Invitrogen™ Supplier No. PA567174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STQACQWSPRA LFHPTGGTQG RRGCRSLLY Highest antigen sequence identity to the following orthologs - mouse 100%, rat 40%.

CYFIP2 (Cytoplasmic FMR1-interacting protein 2, p53-inducible protein 121) is a 1,278 amino acid protein encoded by the human gene CYFIP2. CYFIP2 belongs to the CYFIP family and is involved in T-cell adhesion and p53-dependent induction of apoptosis. It interacts with FMR1, FXR1 and FXR2 and is a component of the WAVE1 complex composed of Abi-2, CYFIP2, C3orf10/HSPC300, NAP125 and WASF1/WAVE1. Upon binding to activated Rac 1, CYFIP2 causes the complex to dissociate, releasing activated WASF1. The CYFIP2 promoter contains a p53-responsive element that confers p53 binding as well as transcriptional activation of a heterologous reporter. Induced expression of CYFIP2 is sufficient for caspase activation and cellular apoptosis, reminiscent of p53 activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYFIP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYFIP2
Gene Accession No. Q96F07
Gene Alias 1500004I01Rik; 6430511D02Rik; AA930218; AU022376; cyfip2; cytoplasmic FMR1 interacting protein 2; cytoplasmic FMR1-interacting protein 2; Kiaa1168; mKIAA1168; p53-inducible protein 121; PIR121
Gene Symbols CYFIP2
Host Species Rabbit
Immunogen Recombinant Human CYFIP2. Recombinant protein control fragment (Product #RP-107817).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26999
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.