Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562823
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562823 Supplier Invitrogen™ Supplier No. PA562823
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASSPFPSSQC TNGHLMCAGC FIHLLADARL KEEQATCPNC RCEISKSLCC RNLAVEKAVS ELPSECGFCL RQFPRSLLER HQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

CYHR1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYHR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CYHR1
Gene Accession No. P0DTL6
Gene Alias 1110031M01Rik; AU042374; Chrp; CYHR1; cysteine and histidine rich 1; cysteine and histidine rich protein; cysteine and histidine-rich cytoplasmic protein; cysteine and histidine-rich protein 1; cysteine/histidine-rich 1; KIAA0496; Unknown (protein for MGC:128912)
Gene Symbols CYHR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CYHR1. Recombinant protein control fragment (Product #RP-93958).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.