Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568516 Supplier Invitrogen™ Supplier No. PA568516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

CYHR1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleoplasm; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYHR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CYHR1
Gene Accession No. 0
Gene Alias 1110031M01Rik; AU042374; Chrp; CYHR1; cysteine and histidine rich 1; cysteine and histidine rich protein; cysteine and histidine-rich cytoplasmic protein; cysteine and histidine-rich protein 1; cysteine/histidine-rich 1; KIAA0496; Unknown (protein for MGC:128912)
Gene Symbols CYHR1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse CYHR1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54151
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.