Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYLC1 Monoclonal Antibody (6F12)
GREENER_CHOICE

Catalog No. PIMA524400
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524400 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524400 Supplier Invitrogen™ Supplier No. MA524400
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SSKTGFKTST KIKGSDTESE ESLYKPGAKK KIDESDGTSA NSKMEGLESK RGFRMSSKKT TFNEKGEKAS TGRVPPSREK PPLPACEPSL PSPKVRRLCW.

Possible architectural role during spermatogenesis. May be involved in spermatid differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYLC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6F12
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CYLC1
Gene Accession No. P35663
Gene Alias CYCL1; CYL; CYL1; CYLC1; cylicin; cylicin 1; cylicin I; cylicin, basic protein of sperm head cytoskeleton 1; cylicin-1; Multiple-band polypeptide I; rCG38076-like
Gene Symbols CYLC1
Host Species Mouse
Immunogen CYLC1 (XP_088636.6, 526 a.a. ~ 625 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1538
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.