Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CYLC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100728 Supplier Invitrogen™ Supplier No. PA5100728
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CYLC2.

Possible architectural role during spermatogenesis. May be involved in spermatid differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYLC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYLC2
Gene Accession No. Q14093
Gene Alias 4930488P18Rik; CYL2; CYLC2; CYLC2 protein; cylicin 2; cylicin II; cylicin, basic protein of sperm head cytoskeleton 2; cylicin-2; LOC100062753; Multiple-band polypeptide II; RGD1562799
Gene Symbols CYLC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYLC2(Accession Q14093), corresponding to amino acid residues T297-G347.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1539, 500458, 74914
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.