Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYLD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Specifications
Antigen | CYLD |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11095574
![]() |
Novus Biologicals
NB11095574 |
0.1 mL | N/A | N/A | N/A | ||||
Description
CYLD Polyclonal specifically detects CYLD in Human, Mouse, Bovine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CYLD | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Cycle and Replication | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
CDMT, CYLD1MFT, CYLDI, cylindromatosis (turban tumor syndrome), Deubiquitinating enzyme CYLD, EAC, EC 3.1.2.15, EC 3.4.19.12, FLJ20180, FLJ31664, KIAA0849FLJ78684, MFT1, probable ubiquitin carboxyl-terminal hydrolase CYLD, SBS, TEM, ubiquitin carboxyl-terminal hydrolase CYLD, ubiquitin specific peptidase like 2, ubiquitin thioesterase CYLD, Ubiquitin thiolesterase CYLD, Ubiquitin-specific-processing protease CYLD, USPL2 | |
CYLD | |
IgG | |
Affinity Purified | |
Human, bovine, orangutan and mouse. Does not react with rat. |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Bovine, Primate, Rat (Negative) | |
Q9NQC7 | |
1540 | |
Synthetic peptide made to an internal portion of human CYLD (within residues 500-600). [Swiss-Prot# Q9NQC7] | |
Primary | |
Store at 4C. Do not freeze. | |
120 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title