Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CYLN2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178744JF549
Description
CYLN2 Polyclonal antibody specifically detects CYLN2 in Human samples. It is validated for Western BlotSpecifications
CYLN2 | |
Polyclonal | |
Western Blot | |
CAP-GLY domain containing linker protein 2, CAP-Gly domain-containing linker protein 2, CLIP, CLIP-115Williams-Beuren syndrome chromosomal region 3 protein, CYLN2WBSCR3, cytoplasmic linker 2, Cytoplasmic linker protein 115, KIAA0291Williams-Beuren syndrome chromosomal region 4 protein, WBSCR4MGC11333, Williams-Beuren syndrome chromosome region 3, Williams-Beuren syndrome chromosome region 4, WSCR3, WSCR4Cytoplasmic linker protein 2 | |
The immunogen for this product maps to a region between residue 996 and 1046 of human Cytoplasmic Linker Protein 115 using the numbering given in entry NP_003379.3 (GeneID 7461). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7461 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction