Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CYM 5541
GREENER_CHOICE

Catalog No. 489710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-971-0 10 mg
48-975-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-971-0 Supplier Tocris Bioscience™ Supplier No. 4897/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective S1P3 receptor allosteric agonist

Selective sphingosine-1-phosphate receptor 3 (S1P3) allosteric agonist (EC50 = 72-132 nM). Occupies a different space within the ligand binding pocket of S1P3 than S1P. Displays no activity at S1P1, S1P2, S1P4 and S1P5 receptors at concentrations up to 10 μM. Shows no significant activities in a profiling panel of 55 GPCRs, ion channels and transporters.

Chemical Identifiers

CAS 945128-26-7
Molecular Formula C19H28N2O2

Specifications

Quantity 10 mg
Formula Weight 316.44
Purity >97%
Molecular Weight (g/mol) 316.445
Chemical Name or Material CYM 5541
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.