Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cymoxanil 98.0+%, TCI America™
SDP

Catalog No. C265725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C265725G 25 g
C26575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C265725G Supplier TCI America Supplier No. C265725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57966-95-7
Molecular Formula C7H10N4O3
Molecular Weight (g/mol) 198.182
MDL Number MFCD00137381
InChI Key XERJKGMBORTKEO-VZUCSPMQSA-N
Synonym 1-(2-Cyano-2-methoxyiminoacetyl)-3-ethylurea, 2-Cyano-N-(ethylcarbamoyl)-2-methoxyiminoacetamide
PubChem CID 5364079
IUPAC Name (1E)-2-(ethylcarbamoylamino)-N-methoxy-2-oxoethanimidoyl cyanide
SMILES CCNC(=O)NC(=O)C(=NOC)C#N

Specifications

Melting Point 162°C
Quantity 25 g
UN Number 3077
Formula Weight 198.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cymoxanil
RTECSNumber AB5957000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.